8-Nitroquinoline 98%

Supplier: Thermo Scientific Chemicals
Warning

B23574-22 B23574-06 B23574-14
AAB23574-22EA 443 USD
AAB23574-22 AAB23574-06 AAB23574-14
8-Nitroquinoline 98%
8-Nitroquinoline

Purity: 98% CAS No.: 607-35-2 Molecular Formula: C9H6N2O2 Formula Weight: 174.16 Melting Point: 86-90° Beilstein Reference: 135178

Formula: C₉H₆N₂O₂
MW: 174.16 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 89 - 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00006806
CAS Number: 607-35-2
EINECS: 210-135-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR