2',4'-Dihydroxypropiophenone 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,4-Dihydroxypropiophenone

B23414-14 B23414-22
AAB23414-14EA 54.4 USD
AAB23414-14 AAB23414-22
2',4'-Dihydroxypropiophenone 99%
2',4'-Dihydroxypropiophenone

Purity: 99% CAS No.: 5792-36-9 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 98-101° Beilstein Reference: 1238635

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 343 °C (1013 hPa)
Melting Pt: 101.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00002280
CAS Number: 5792-36-9
EINECS: 227-329-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR