2-(Aminomethyl)pyridine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Pyridylmethylamine, 2-Picolylamine, Pyridin-2-ylmethanamine, (2-Pyridyl)methylamine, 2-Picolamine
B23208-14
B23208-06
B23208-22
AAB23208-14EA
80.9
USD
AAB23208-14
AAB23208-06
AAB23208-22
2-(Aminomethyl)pyridine 98+%
2-(Aminomethyl)pyridine
Purity: 98+%
CAS No.: 3731-51-9
Molecular Formula: C6H8N2
Formula Weight: 108.14
Flash Point: 90°(194°F)
Refractive Index: 1.5450
Odor: Amine-like
pH: 11-12
Melting Point: -20°
Boiling Point: 202-204°
Beilstein Reference: 108054
UN No.: 2735
MDL No.: MFCD00006360
Formula:
C₆H₈N₂ Boiling Pt: 202…204 °C Melting Pt: –20 °C Density: 1.0583 g/cm³ (20 °C) Flash Pt: 90 °C (194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006360 CAS Number: 3731-51-9 EINECS: 223-090-5 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...