4-Methoxybenzophenone 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: p-Methoxybenzophenone, (4-Methoxyphenyl)(phenyl)methanone

B23123-14 B23123-22
AAB23123-14EA 62.5 USD
AAB23123-14 AAB23123-22
4-Methoxybenzophenone 98+%
4-Methoxybenzophenone

Purity: 98+% CAS No.: 611-94-9 Molecular Formula: C14H12O2 Formula Weight: 212.25 Melting Point: 60-63° Boiling Point: 354-356° MDL No.: MFCD00008403

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 354…356 °C
Melting Pt: 60…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00008403
CAS Number: 611-94-9
EINECS: 210-285-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR