4-Methyl-m-anisic acid 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-Methoxy-4-methylbenzoic acid

B22982-06 B22982-14
AAB22982-03EA 35.3 USD
AAB22982-03 AAB22982-06 AAB22982-14
4-Methyl-m-anisic acid 99%
4-Methyl-m-anisic acid

Purity: 99% CAS No.: 7151-68-0 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 158-160° Beilstein Reference: 1938389

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Melting Pt: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00002566
CAS Number: 7151-68-0
EINECS: 230-486-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR