Phenyl acetate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Purity: 99%
CAS No.: 122-79-2
Molecular Formula: CH3CO2C6H5
Formula Weight: 136.15
Flash Point: 76°(168°F)
Refractive Index: 1.5020
Odor: Phenol-like
Boiling Point: 195-196°
Beilstein Reference: 636458
Merck Reference: 14,7267
MDL No.: MFCD00008699
Formula:
C₈H₈O₂ Boiling Pt: 195…196 °C Density: 1.075 Flash Pt: 76 °C(168 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008699 CAS Number: 122-79-2 EINECS: 204-575-0 Merck Index: 14,7267 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...