1-Bromoperfluorooctane 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Purity: 98+%
CAS No.: 423-55-2
Molecular Formula: C8H17Br
Formula Weight: 498.98
Flash Point: None
Refractive Index: 1.3050
Melting Point: 6°
Boiling Point: 141-143°
Beilstein Reference: 2016022
Merck Reference: 14,7156
MDL No.: MFCD00042082
Formula:
BrF₂C(CF₂)₆CF₃ Boiling Pt: 141…143 °C Melting Pt: 6 °C Density: 1.930 Storage Temperature: Ambient |
MDL Number:
MFCD00042082 CAS Number: 423-55-2 EINECS: 207-028-4 Merck Index: 14,7156 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...