Dimethyl pimelate 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: Dimethyl heptanedioate, 1,7-Heptanedioic acid dimethyl ester, Pimelic acid dimethyl ester

B22026-06 B22026-14 B22026-22
AAB22026-06EA 41.9 USD
AAB22026-06 AAB22026-14 AAB22026-22
Dimethyl pimelate 98+%
Dimethyl pimelate

Purity: 99% CAS No.: 1732-08-7 Molecular Formula: C9H16O4 Formula Weight: 188.23 Flash Point: 110°(230°F) Refractive Index: 1.4310 Odor: Mild Boiling Point: 121-122°/11mm Beilstein Reference: 1777309 MDL No.: MFCD00008470

Formula: C₉H₁₆O₄
MW: 188.22 g/mol
Boiling Pt: 121…122 °C/11 mm
Melting Pt: -18.7 °C
Density: 1.041 g/cm³ (20 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008470
CAS Number: 1732-08-7
EINECS: 217-057-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR