1,1'-Thiocarbonyldiimidazole tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

B22008-30 B22008-09 B22008-18
AAB22008-30EA 1268 USD
AAB22008-30 AAB22008-09 AAB22008-18
1,1'-Thiocarbonyldiimidazole tech. 90%, Technical Grade
1,1'-Thiocarbonyldiimidazole

Purity: 90% CAS No.: 6160-65-2 Molecular Formula: C7H6N4S Formula Weight: 178.22 Storage Temperature: 2°C to 8°C Melting Point: 98-102° Beilstein Reference: 609349 MDL No.: MFCD00005289

Formula: C₇H₆N₄S
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00005289
CAS Number: 6160-65-2
EINECS: 228-183-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR