(R)-(-)-1,2-Propanediol 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(R)-(-)-1,2-Dihydroxy propane, (R)-(-)-1,2-Propylene glykol, (R)-Propane-1,2-diol
B21937-03
B21937-06
B21937-14
AAB21937-03EA
45.6
USD
AAB21937-03
AAB21937-06
AAB21937-14
(R)-(-)-1,2-Propanediol 98%
(R)-(-)-1,2-Propanediol
Purity: 98%
CAS No.: 4254-14-2
Molecular Formula: C3H8O2
Formula Weight: 76.10
Flash Point: 107°(224°F)
Refractive Index: 1.4320
Boiling Point: 186-188°
Beilstein Reference: 1718872
Merck Reference: 14,7855
Optical Rotation: -16.5° (neat)
MDL No.: MFCD00066248
Formula:
C₃H₈O₂ MW: 76.1 g/mol Boiling Pt: 186…188 °C Density: 1.040 Flash Pt: 107 °C(224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00066248 CAS Number: 4254-14-2 Merck Index: 14,7855 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...