6-Chloronicotinamide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Chloropyridine-3-carboxamide

B21922-14 B21922-03 B21922-06
AAB21922-14EA 213 USD
AAB21922-14 AAB21922-03 AAB21922-06
6-Chloronicotinamide 98%
6-Chloronicotinamide

Purity: 98% CAS No.: 6271-78-9 Molecular Formula: C6H5ClN2O Formula Weight: 156.57 Melting Point: 214-216° Beilstein Reference: 116043

Formula: C₆H₅ClN₂O
MW: 156.57 g/mol
Melting Pt: 214…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00006242
CAS Number: 6271-78-9
EINECS: 228-456-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR