2-Bromobiphenyl (PBB No. 1) 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromobiphenyl, PBB No. 1, 2-Bromodiphenyl

B21840-14 B21840-03 B21840-06
AAB21840-14EA 279 USD
AAB21840-14 AAB21840-03 AAB21840-06
2-Bromobiphenyl (PBB No. 1) 98%
2-Bromobiphenyl (PBB No. 1)

Purity: 98% CAS No.: 2052-07-5 Molecular Formula: C12H9Br Formula Weight: 233.12 Flash Point: 110°(230°F) Refractive Index: 1.6290 Melting Point: 0-2° Boiling Point: 298° Beilstein Reference: 1863156 MDL No.: MFCD00000065

Formula: C₁₂H₉Br
MW: 233.11 g/mol
Boiling Pt: 298 °C
Melting Pt: 0…2 °C
Density: 1.2766 g/cm³ (20 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000065
CAS Number: 2052-07-5
EINECS: 218-141-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR