6-Hydroxy-2-naphthoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-hydroxynaphthalene-2-carboxylic acid
B21791-14
B21791-06
AAB21791-14EA
226
USD
AAB21791-14
AAB21791-03
AAB21791-06
6-Hydroxy-2-naphthoic acid 99%
6-Hydroxy-2-naphthoic acid
Purity: 99%
CAS No.: 16712-64-4
Molecular Formula: C11H8O3
Formula Weight: 188.18
Melting Point: 248-250°
Beilstein Reference: 2692084
Formula:
C₁₁H₈O₃ Melting Pt: 248…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060070 CAS Number: 16712-64-4 EINECS: 240-759-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...