5-Fluorosalicylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Fluoro-2-hydroxybenzoic acid

B21751-06 B21751-03
AAAB21751-06EA 211 USD
AAAB21751-06 AAAB21751-MD AAB21751-03
5-Fluorosalicylic acid 97%
5-Fluorosalicylic acid

Purity: 97% CAS No.: 345-16-4 Molecular Formula: C7H5FO3 Formula Weight: 156.11 Melting Point: 178-182° Beilstein Reference: 2209120

Formula: C₇H₅FO₃
MW: 156.11 g/mol
Boiling Pt: 295
Melting Pt: 178…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00002456
CAS Number: 345-16-4
EINECS: 206-455-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR