3-Bromobiphenyl (PBB No. 2) 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
PBB No. 2
B21726-06
B21726-14
B21726-03
AAB21726-06EA
157
USD
AAB21726-06
AAB21726-14
AAB21726-03
3-Bromobiphenyl (PBB No. 2) 99%
3-Bromobiphenyl (PBB No. 2)
Purity: 99%
CAS No.: 2113-57-7
Molecular Formula: C12H9Br
Formula Weight: 233.12
Flash Point:
Formula:
C₁₂H₉Br MW: 233.11 g/mol Boiling Pt: 301 °C Melting Pt: 9 °C Density: 1.41 g/cm³ (20 °C) Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000082 CAS Number: 2113-57-7 EINECS: 218-304-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...