Octafluoronaphthalene 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Perfluoronaphthalene

B21699-06 B21699-14 B21699-03
AAB21699-06EA 134 USD
AAB21699-06 AAAB21699-14 AAB21699-03
Octafluoronaphthalene 96%
Octafluoronaphthalene

Purity: 96% CAS No.: 313-72-4 Molecular Formula: C10F8 Formula Weight: 272.10 Melting Point: 84-89° Boiling Point: 209° MDL No.: MFCD00014307

Formula: C₁₀F₈
MW: 272.1 g/mol
Boiling Pt: 238 °C (1013 hPa)
Melting Pt: 80 - 86 °C
Density: 2.069 g/cm³ (-70 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014307
CAS Number: 313-72-4
EINECS: 206-239-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR