4'-Methylpropiophenone 94%
Supplier: Thermo Scientific Chemicals
Synonyms:
p-Methylpropiophenone
B21673-36
B21673-22
AAB21673-36EA
127
USD
AAB21673-36
AAB21673-22
4'-Methylpropiophenone 94%
4'-Methylpropiophenone
Purity: 94%
CAS No.: 5337-93-9
Molecular Formula: C10H12O
Formula Weight: 148.21
Flash Point: 108°(226°F)
Refractive Index: 1.5280
Melting Point: 7°
Boiling Point: 238-239°
Beilstein Reference: 2042137
MDL No.: MFCD00009312
Formula:
C₁₀H₁₂O MW: 148.2 g/mol Boiling Pt: 238…239 °C Melting Pt: 7 °C Density: 1.0053 g/cm³ (0 °C) Flash Pt: 108 °C(226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009312 CAS Number: 5337-93-9 EINECS: 226-267-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...