2'-Acetoxyacetophenone 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-Acetylphenyl acetate, 2′-Acetoxyacetophenone, o-Acetylphenyl acetate
B21624-14
B21624-06
AAB21624-14EA
109
USD
AAB21624-14
AAB21624-06
AAB21624-22
2'-Acetoxyacetophenone 98%
2'-Acetoxyacetophenone
Purity: 98%
CAS No.: 7250-94-4
Molecular Formula: C10H10O3
Formula Weight: 178.19
Melting Point: 88-91°
Boiling Point: 118°/19mm
MDL No.: MFCD00017223
Formula:
C₁₀H₁₀O₃ Boiling Pt: 118 °C/19 mm Melting Pt: 88…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017223 CAS Number: 7250-94-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...