2-Fluoro-4-hydroxybenzonitrile 99%

Supplier: Thermo Scientific Chemicals
Danger

B21535-03 B21535-06
AAB21535-03EA 34.7 USD
AAB21535-03 AAB21535-06 AAB21535-14
2-Fluoro-4-hydroxybenzonitrile 99%
2-Fluoro-4-hydroxybenzonitrile

Purity: 99% CAS No.: 82380-18-5 Molecular Formula: C7H4FNO Formula Weight: 137.11 Melting Point: 123-126° Beilstein Reference: 4741066 UN No.: 3077 MDL No.: MFCD00192177

Formula: C₇H₄FNO
Boiling Pt: 261 °C (1013 hPa)
Melting Pt: 123…126 °C
Density: 1.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00192177
CAS Number: 82380-18-5
EINECS: 422-810-7
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR