(±)-3-Quinuclidinol 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (±)-Quinuclidin-3-ol, (±)-3-Hydroxyquinuclidine

B21503-06 B21503-14 B21503-22
AAB21503-06EA 42.02 USD
AAB21503-06 AAB21503-14 AAB21503-22
(±)-3-Quinuclidinol 98+%
(±)-3-Quinuclidinol

Purity: 98+% CAS No.: 1619-34-7 Molecular Formula: C7H13NO Formula Weight: 127.19 pH: 10-12 Melting Point: 217-224° Beilstein Reference: 104327 Merck Reference: 14,8082 UN No.: 3263 MDL No.: MFCD00151326

Formula: C₇H₁₃NO
MW: 127.19 g/mol
Melting Pt: 217…224 °C
Storage Temperature: Ambient
MDL Number: MFCD00151326
CAS Number: 1619-34-7
EINECS: 216-578-4
UN: 3259
ADR: 8,III
Merck Index: 12,08266

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR