3-Hydroxy-p-toluic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Hydroxy-4-methylbenzoic acid

B21501-03 B21501-14 B21501-06
AAB21501-03EA 17 USD
AAB21501-03 AAB21501-14 AAB21501-06
3-Hydroxy-p-toluic acid 98%
3-Hydroxy-p-toluic acid

Purity: 98% CAS No.: 586-30-1 Molecular Formula: C8H8O3 Formula Weight: 152.15 Odor: Odorless Melting Point: 207-212° Beilstein Reference: 1936497 MDL No.: MFCD00002511

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 207…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00002511
CAS Number: 586-30-1
EINECS: 209-571-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR