Propylbenzene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21468-22
B21468-36
AAB21468-22EA
61.4
USD
AAB21468-22
AAB21468-36
Propylbenzene 98%
Propylbenzene
Purity: 98%
CAS No.: 103-65-1
Molecular Formula: C9H12
Formula Weight: 120.20
Flash Point: 47°(116°F)
Refractive Index: 1.4910
Melting Point: -100°
Boiling Point: 159-160°
Beilstein Reference: 1903006
Merck Reference: 14,7844
UN No.: 1993
MDL No.: MFCD00009377
Formula:
C₉H₁₂ MW: 120.19 g/mol Boiling Pt: 159…160 °C Melting Pt: –100 °C Density: 0.862 g/cm³ (20 °C) Flash Pt: 47 °C (116 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009377 CAS Number: 103-65-1 EINECS: 203-132-9 UN: 2364 ADR: 3,III Merck Index: 12,08029 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...