4-Bromo-3,5-dimethoxybenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B21449-06 B21449-14
AAB21449-06EA 45.3 USD
AAB21449-06 AAB21449-14
4-Bromo-3,5-dimethoxybenzoic acid 98%
4-Bromo-3,5-dimethoxybenzoic acid

Purity: 98% CAS No.: 56518-42-4 Molecular Formula: C9H9BrO4 Formula Weight: 261.07 Melting Point: 254-258° MDL No.: MFCD01632140

Formula: C₉H₉BrO₄
MW: 261.07 g/mol
Melting Pt: 254…258 °C
Storage Temperature: Ambient
MDL Number: MFCD01632140
CAS Number: 56518-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR