1,3-Dimethylurea 98%

Supplier: Thermo Scientific Chemicals

Synonyms: N,N'-Dimethylurea

B21329-30 B21329-0B B21329-36
AAB21329-30EA 30.26 USD
AAB21329-30 AAB21329-0B AAAB21329-36
1,3-Dimethylurea 98%
1,3-Dimethylurea

Purity: 98% CAS No.: 96-31-1 Molecular Formula: (CH3NH)2C=O Formula Weight: 88.11 Flash Point: 157°(314°F) Melting Point: 100-104° Boiling Point: 268-270° dec. Beilstein Reference: 1740672 MDL No.: MFCD00008286

Formula: C₃H₈N₂O
MW: 88.11 g/mol
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 96 °C
Density: 1.142 g/cm³ (20 °C)
Flash Pt: 157 °C(314 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008286
CAS Number: 96-31-1
EINECS: 202-498-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR