CAPSO (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid) 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-(Cyclohexylamino)-2-hydroxy-1-propanesulphonic acid

B21305-22 B21305-14 B21305-36
AAB21305-22EA 119.57 USD
AAB21305-22 AAB21305-14 AAB21305-36
CAPSO (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid) 98%
CAPSO (3-(cyclohexylamino)-2-hydroxypropanesulphonic acid)

Purity: 98% CAS No.: 73463-39-5 Molecular Formula: C9H19NO4S Formula Weight: 237.32 Melting Point: 270-274° MDL No.: MFCD00041778

Formula: C₉H₁₉NO₄S
Melting Pt: 270…274 °C
Storage Temperature: Ambient
MDL Number: MFCD00041778
CAS Number: 73463-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR