Tetrabutylammonium fluoride ≥75% (w/w) in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21261-14
B21261-36
B21261-22
AAB21261-14EA
41.1
USD
AAB21261-14
AAB21261-36
AAB21261-22
Tetrabutylammonium fluoride ≥75% (w/w) in aqueous solution
Tetrabutylammonium fluoride
CAS No.: 429-41-4
Molecular Formula: [CH3(CH2)3]4NF
Formula Weight: 261.47
UN No.: 2922
MDL No.: MFCD00011747
Formula:
C₁₆H₃₆FN MW: 261.47 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011747 CAS Number: 429-41-4 EINECS: 207-057-2 UN: 2922 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...