Diphenylmethane 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21256-0E
B21256-22
B21256-36
AAB21256-0EEA
201
USD
AAB21256-0E
AAB21256-22
AAB21256-36
Diphenylmethane 99+%
Diphenylmethane
Purity: 99+%
CAS No.: 101-81-5
Molecular Formula: (C6H5)2CH2
Formula Weight: 168.24
Flash Point: 130°(266°F)
Refractive Index: 1.5770
Melting Point: 22-26°
Boiling Point: 262-264°
Beilstein Reference: 1904982
Merck Reference: 14,3328
UN No.: 3077
MDL No.: MFCD00004781
Formula:
C₁₃H₁₂ MW: 168.24 g/mol Boiling Pt: 262…264 °C Melting Pt: 22…26 °C Density: 0.9985 g/cm³ (20 °C) Flash Pt: 130 °C(266 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004781 CAS Number: 101-81-5 EINECS: 202-978-6 UN: 3077 ADR: 9,III Merck Index: 13,03360 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...