1,12-Dibromododecane 98%

Supplier: Thermo Scientific Chemicals

B21254-22 B21254-06 B21254-14
AAAB21254-22EA 294 USD
AAAB21254-22 AAB21254-06 AAB21254-14
1,12-Dibromododecane 98%
1,12-Dibromododecane

Purity: 98% CAS No.: 3344-70-5 Molecular Formula: Br(CH2)12Br Formula Weight: 328.13 Flash Point: 110°(230°F) Melting Point: 38-42° Boiling Point: 215°/15mm Beilstein Reference: 1742763 MDL No.: MFCD00000226

Formula: BrH₂C(CH₂)₁₀CH₂Br
MW: 328.13 g/mol
Boiling Pt: 215 °C/15 mm
Melting Pt: 38…42 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000226
CAS Number: 3344-70-5
EINECS: 222-096-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR