6-Fluoroveratraldehyde 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Fluoro-4,5-dimethoxybenzaldehyde, 4,5-Dimethoxy-2-fluorobenzaldehyde, 3,4-Dimethoxy-6-fluorobenzaldehyde
B21202-14
B21202-03
B21202-06
AAAB21202-14EA
716
USD
AAAB21202-14
AAB21202-03
AAB21202-06
6-Fluoroveratraldehyde 98+%
6-Fluoroveratraldehyde
Purity: 98+%
CAS No.: 71924-62-4
Molecular Formula: C9H9FO3
Formula Weight: 184.17
Melting Point: 98-100°
Beilstein Reference: 2109343
Formula:
C₉H₉FO₃ MW: 184.17 g/mol Melting Pt: 98…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061108 CAS Number: 71924-62-4 EINECS: 676-950-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...