Resazurin sodium salt
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
7-Sodiooxy-3H-phenoxazin-3-one 10-oxide, Sodium 7-oxido-3-oxo-3H-phenoxazine 10-oxide, Resazurin sodium, Resazurin
B21187-06
B21187-03
AAB21187-06EA
132.8
USD
AAB21187-06
AAB21187-03
Resazurin sodium salt
Resazurin sodium salt
As indicator pH 3.8 (orange) to 6.5 (dark violet)
CAS No.: 62758-13-8
Molecular Formula: C12H6NNaO4
Formula Weight: 251.18
Physical Form: Powder
Odor: Odorless
Beilstein Reference: 3794971
Merck Reference: 14,8141
MDL No.: MFCD00005036
As indicator pH 3.8 (orange) to 6.5 (dark violet)
Formula:
C₁₂H₆NNaO₄ MW: 251.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005036 CAS Number: 62758-13-8 EINECS: 263-718-5 Merck Index: 14,8141 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...