Dimethyl adipate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Adipic acid dimethyl ester, 1,6-Hexanedioic acid dimethyl ester
B21174-30
B21174-18
B21174-0B
AAB21174-30EA
34.8
USD
AAB21174-30
AAAB21174-18
AAB21174-0B
Dimethyl adipate 99%
Dimethyl adipate
Purity: 99%
CAS No.: 627-93-0
Molecular Formula: CH3O2C(CH2)4CO2CH3
Formula Weight: 174.20
Flash Point: 107°(224°F)
Refractive Index: 1.4280
Melting Point: 8°
Boiling Point: 109-110°/14mm
Beilstein Reference: 1707443
Merck Reference: 14,162
MDL No.: MFCD00008469
Formula:
C₈H₁₄O₄ MW: 174.2 g/mol Boiling Pt: 229 Melting Pt: 10.3 Density: 1.063 Flash Pt: 107 °C(224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008469 CAS Number: 627-93-0 EINECS: 211-020-6 Merck Index: 14,162 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...