2,4-Dinitroaniline 99% (dry weight) wetted with ca. 15% water
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21080-22
B21080-36
AAB21080-0EEA
208.8
USD
AAB21080-0E
AAB21080-22
AAB21080-36
2,4-Dinitroaniline 99% (dry weight) wetted with ca. 15% water
2,4-Dinitroaniline
Purity: 99%
CAS No.: 97-02-9
Molecular Formula: C6H5N3O4
Formula Weight: 183.12
Flash Point: 224°(435°F)
Melting Point: 176-180°
Beilstein Reference: 982999
Merck Reference: 14,3270
UN No.: 1596
MDL No.: MFCD00007151
Formula:
C₆H₅N₃O₄ Boiling Pt: 401 °C (1013 hPa) Melting Pt: 176…180 °C Density: 1.61 Flash Pt: 224 °C (435 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007151 CAS Number: 97-02-9 EINECS: 202-553-5 UN: 1596 ADR: 6.1,II Merck Index: 14,3270 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...