2-Ethyl-1-butanol 99%

Supplier: Thermo Scientific Chemicals
Warning

B21026-0F B21026-AP
AAB21026-0FEA 1598 USD
AAB21026-0F AAB21026-AP
2-Ethyl-1-butanol 99%
2-Ethyl-1-butanol

Purity: 99% CAS No.: 97-95-0 Molecular Formula: (CH3CH2)2CHCH2OH Formula Weight: 102.18 Flash Point: 58°(136°F) Refractive Index: 1.4220 Melting Point: -52° Boiling Point: 146-148° Beilstein Reference: 1731254 UN No.: 1987 MDL No.: MFCD00004744

Formula: C₆H₁₄O
Boiling Pt: 146…148 °C
Melting Pt: –52 °C
Density: 0.8233 g/cm³ (20 °C)
Flash Pt: 58 °C (136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004744
CAS Number: 97-95-0
EINECS: 202-621-4
UN: 2275
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR