3-Methoxyphenylacetone 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(3-Methoxyphenyl)propan-2-one

B20916-06 B20916-03
AAB20916-06EA 323 USD
AAB20916-06 AAB20916-03
3-Methoxyphenylacetone 97%
3-Methoxyphenylacetone

Purity: 97% CAS No.: 3027-13-2 Molecular Formula: C10H12O2 Formula Weight: 164.20 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.078 Refractive Index: 1.5252 Appearance: Pale yellow liquid Boiling Point: 258-260° Beilstein Reference: 2044357 MDL No.: MFCD00008771

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 258…260 °C
Density: 1.078
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008771
CAS Number: 3027-13-2
EINECS: 221-191-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR