1-Phenylcyclohexanol 97%
Supplier: Thermo Scientific Chemicals
B20877-18
B20877-09
AAB20877-18EA
199
USD
AAB20877-18
AAB20877-09
1-Phenylcyclohexanol 97%
1-Phenylcyclohexanol
Purity: 97%
CAS No.: 1589-60-2
Molecular Formula: C12H16O
Formula Weight: 176.26
Melting Point: 58-62°
Boiling Point: 153°/20mm
MDL No.: MFCD00021393
Formula:
C₁₂H₁₆O Boiling Pt: 153 °C/20 mm Melting Pt: 58…62 °C Density: 1.101 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00021393 CAS Number: 1589-60-2 EINECS: 216-456-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...