1-Amino-2,2,2-trifluoroethane 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,2,2-Trifluoroethylamine

B20789-06 B20789-03 B20789-14
AAB20789-06EA 149 USD
AAB20789-06 AAB20789-03 AAB20789-14
1-Amino-2,2,2-trifluoroethane 98%
1-Amino-2,2,2-trifluoroethane

Purity: 98% CAS No.: 753-90-2 Molecular Formula: CF3CH2NH2 Formula Weight: 99.06 Storage Temperature: 2°C to 8°C Flash Point: -16°(3°F) Refractive Index: 1.2950 Odor: Amine-like Boiling Point: 36-38° Beilstein Reference: 1733204 UN No.: 2733 MDL No.: MFCD00008132

Formula: C₂H₄F₃N
MW: 99.06 g/mol
Boiling Pt: 36…38 °C
Density: 1.2452 g/cm³ (25 °C)
Flash Pt: –16 °C (3 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008132
CAS Number: 753-90-2
EINECS: 212-041-3
UN: 3286
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR