3-Cyclohexylpropyne 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Propynyl)cyclohexane, (2-Propinyl)cyclohexan, Prop-2-yn-1-ylcyclohexane

B20505-06 B20505-03
AAB20505-06EA 148 USD
AAB20505-06 AAB20505-03 AAB20505-14
3-Cyclohexylpropyne 97%
3-Cyclohexylpropyne

Purity: 97% CAS No.: 17715-00-3 Molecular Formula: C9H14 Formula Weight: 122.21 Flash Point: 35°(95°F) Refractive Index: 1.4620 Boiling Point: 157-158° Beilstein Reference: 1849402 UN No.: 3295 MDL No.: MFCD00041562

Formula: C₉H₁₄
MW: 122.21
Boiling Pt: 48
Density: 0.845
Flash Pt: 35 °C (95 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041562
CAS Number: 17715-00-3
UN: 3295
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR