8-Chloroquinaldine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 8-Chloro-2-methylquinoline

B20504-03 B20504-06
AAB20504-03EA 29.8 USD
AAB20504-03 AAB20504-06
8-Chloroquinaldine 98%
8-Chloroquinaldine

Purity: 98% CAS No.: 3033-82-7 Molecular Formula: C10H8ClN Formula Weight: 177.63 Melting Point: 65-68° Beilstein Reference: 119193

Formula: C₁₀H₈ClN
MW: 177.63 g/mol
Melting Pt: 65…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00051764
CAS Number: 3033-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR