Ursodeoxycholic acid 99%
Supplier: Thermo Scientific Chemicals
B20490-06
B20490-14
B20490-03
AAB20490-06EA
126
USD
AAB20490-06
AAB20490-14
AAB20490-03
Ursodeoxycholic acid 99%
Ursodeoxycholic acid
A secondary bile acid
Purity: 99%
CAS No.: 128-13-2
Molecular Formula: C24H40O4
Formula Weight: 392.56
Melting Point: 201-204°
Merck Reference: 14,9889
MDL No.: MFCD00003680
Formula:
C₂₄H₄₀O₄ MW: 392.58 g/mol Boiling Pt: 547 °C (1013 hPa) Melting Pt: 201…204 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003680 CAS Number: 128-13-2 EINECS: 204-879-3 Merck Index: 12,10026 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...