Ethyl bromodifluoroacetate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethyl 2-bromo-2,2-difluoroacetate, Bromodifluoroacetic ethyl ester

B20461-06 B20461-14
AAB20461-06EA 68.9 USD
AAB20461-06 AAB20461-14
Ethyl bromodifluoroacetate 97%
Ethyl bromodifluoroacetate

Purity: 97% CAS No.: 667-27-6 Molecular Formula: BrCF2CO2CH2CH3 Formula Weight: 202.99 Flash Point: 21°(69°F) Refractive Index: 1.3880 Boiling Point: 112-114° Beilstein Reference: 1906095 UN No.: 2924 MDL No.: MFCD00042069

Formula: BrF₂CCOOCH₂CH₃
MW: 202.98 g/mol
Boiling Pt: 112…114 °C
Density: 1.5926 g/cm³ (20 °C)
Flash Pt: 21 °C (69 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042069
CAS Number: 667-27-6
EINECS: 211-567-0
UN: 2924
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR