2,4,4-Trimethylcyclopentanone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20425-03
B20425-06
AAB20425-03EA
40.5
USD
AAB20425-03
AAB20425-06
2,4,4-Trimethylcyclopentanone 98%
2,4,4-Trimethylcyclopentanone
Purity: 98%
CAS No.: 4694-12-6
Molecular Formula: C8H14O
Formula Weight: 126.20
Flash Point: 43°(110°F)
Refractive Index: 1.4320
Boiling Point: 159-161°
UN No.: 1224
MDL No.: MFCD00019313
Formula:
C₈H₁₄O MW: 126.2 g/mol Boiling Pt: 159…161 °C Melting Pt: -25.6 °C Density: 0.8772 g/cm³ (20 °C) Flash Pt: 43 °C (110 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00019313 CAS Number: 4694-12-6 EINECS: 225-155-3 UN: 1224 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...