2,3,6-Trifluorophenol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B20379-03
B20379-06
AAB20379-03EA
42.4
USD
AAB20379-03
AAB20379-06
2,3,6-Trifluorophenol 98%
2,3,6-Trifluorophenol
Purity: 98%
CAS No.: 113798-74-6
Molecular Formula: C6H3F3O
Formula Weight: 148.09
Flash Point: 63°(145°F)
Melting Point: 31-36°
Beilstein Reference: 4976927
UN No.: 3261
MDL No.: MFCD00061214
Formula:
C₆H₃F₃O MW: 148.08 g/mol Boiling Pt: 155 °C (1013 hPa) Melting Pt: 31…36 °C Flash Pt: 63 °C (145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00061214 CAS Number: 113798-74-6 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...