1,2-Dibromotetrachloroethane 97%

Supplier: Thermo Scientific Chemicals
Warning

B20276-14 B20276-22
AAB20276-14EA 42.3 USD
AAB20276-14 AAB20276-22
1,2-Dibromotetrachloroethane 97%
1,2-Dibromotetrachloroethane

Purity: 97% CAS No.: 630-25-1 Molecular Formula: BrCCl2CCl2Br Formula Weight: 325.65 Melting Point: ca 220° dec. Beilstein Reference: 1699471

Formula: BrCl₂CCBrCl₂
MW: 325.64 g/mol
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: ∼ 220 °C (dec.)
Density: 2.713 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000780
CAS Number: 630-25-1
EINECS: 211-136-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR