1-Iodoperfluorooctane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Perfluorooctyl iodide, Heptadecafluoro-1-iodooctane, Perfluoro-1-iodooctane
B20207-09
B20207-18
AAB20207-09EA
69.4
USD
AAB20207-09
AAB20207-18
1-Iodoperfluorooctane 98%
1-Iodoperfluorooctane
Purity: 98%
CAS No.: 507-63-1
Molecular Formula: C8F17I
Formula Weight: 545.97
Refractive Index: 1.3290
Melting Point: 25°
Boiling Point: 160-161°
Beilstein Reference: 1717141
MDL No.: MFCD00001064
Formula:
C₈F₁₇I MW: 545.97 g/mol Boiling Pt: 160…161 °C Melting Pt: 25 °C Density: 2.071 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001064 CAS Number: 507-63-1 EINECS: 208-079-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...