14-Heptacosanone 97%

Supplier: Thermo Scientific Chemicals

B20191-06 B20191-14
AAB20191-06EA 102 USD
AAB20191-06 AAB20191-14
14-Heptacosanone 97%
14-Heptacosanone

Purity: 97% CAS No.: 542-50-7 Molecular Formula: C27H54O Formula Weight: 394.73 Melting Point: 76-79° Beilstein Reference: 1796794

Formula: C₂₇H₅₄O
MW: 394.73 g/mol
Melting Pt: 76…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00026598
CAS Number: 542-50-7
EINECS: 208-815-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR