2,4,4-Trimethyl-1-pentene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 99%
CAS No.: 107-39-1
Molecular Formula: C8H16
Formula Weight: 112.22
Flash Point: -6°(21°F)
Refractive Index: 1.4085
Odor: Petrol-like
Melting Point: -94°
Boiling Point: 101-102°
Beilstein Reference: 1098309
UN No.: 2050
MDL No.: MFCD00008855
Formula:
C₈H₁₆ Boiling Pt: 101…102 °C Melting Pt: –94 °C Density: 0.71509 g/cm³ (20 °C) Flash Pt: –6 °C (21 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008855 CAS Number: 107-39-1 EINECS: 203-486-4 UN: 2050 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...