2,4-Dichloroanisole 99%

Supplier: Thermo Scientific Chemicals
Warning

B20042-06 B20042-22 B20042-14
AAB20042-06EA 24.46 USD
AAB20042-06 AAB20042-22 AAB20042-14
2,4-Dichloroanisole 99%
2,4-Dichloroanisole

Purity: 99% CAS No.: 553-82-2 Molecular Formula: C7H6Cl2O Formula Weight: 177.03 Flash Point: 110°(230°F) Refractive Index: 1.5615 Melting Point: 24-27° Boiling Point: 125°/10mm Beilstein Reference: 1940358 MDL No.: MFCD00044772

Formula: C₇H₆Cl₂O
Boiling Pt: 125 °C/10 mm
Melting Pt: 24…27 °C
Density: 1.288
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00044772
CAS Number: 553-82-2
EINECS: 209-051-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR