4-Ethoxybenzonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
p-Ethoxybenzonitrile
A19806-06
A19806-14
A19806-22
AAA19806-06EA
45.8
USD
AAA19806-06
AAA19806-14
AAAA19806-22
4-Ethoxybenzonitrile 98%
4-Ethoxybenzonitrile
Purity: 98%
CAS No.: 25117-74-2
Molecular Formula: C9H9NO
Formula Weight: 147.18
Melting Point: 60-63°
Boiling Point: 258°
UN No.: 3439
MDL No.: MFCD00001819
Formula:
C₉H₉NO MW: 147.18 g/mol Boiling Pt: 258 Melting Pt: 60…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001819 CAS Number: 25117-74-2 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...