3-Aminobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A19540-36 A19540-22 A19540-0E
AAA19540-36EA 138 USD
AAA19540-36 AAA19540-22 AAAA19540-0E
3-Aminobenzoic acid 98%
3-Aminobenzoic acid

Purity: 98% CAS No.: 99-05-8 Molecular Formula: C7H7NO2 Formula Weight: 137.14 Flash Point: 150°(302°F) Melting Point: 172-179° Beilstein Reference: 471603 Merck Reference: 14,421 MDL No.: MFCD00007795

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 172…179 °C
Density: 1.5105 g/cm³ (20 °C)
Flash Pt: 150 °C(302 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007795
CAS Number: 99-05-8
EINECS: 202-724-4
Merck Index: 13,00420

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR