(4-(Trifluoromethoxy)phenyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Trifluoromethoxy)phenylacetic acid

A19512-03 A19512-06
AAA19512-03EA 61.3 USD
AAA19512-03 AAA19512-06
(4-(Trifluoromethoxy)phenyl)acetic acid 98%
(4-(Trifluoromethoxy)phenyl)acetic acid

Purity: 98% CAS No.: 4315-07-5 Molecular Formula: C9H7F3O3 Formula Weight: 220.15 Melting Point: 86-88° Beilstein Reference: 2695336

Formula: (C₆H₄(OCF₃))CH₂COOH
MW: 220.15 g/mol
Melting Pt: 86…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00061239
CAS Number: 4315-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR